ENA394767602 Enamine

2-(4-cyanophenyl)-2-hydroxyacetic acid

  • MDL number MFCD20658797
  • Cas Number : 631897-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128984
Availability details for
Quantity
  • Availability
  • Add to Cart