ENA394767603 Enamine

4-fluoro-1-(2-phenoxyethyl)-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD24842914
  • Cas Number : 1304465-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128985

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-128985
Availability details for
Quantity
  • Availability
  • Add to Cart