ENA394767605 Enamine

[(3-chloro-4-methoxyphenyl)amino]carbonitrile

  • MDL number MFCD16767915
  • Cas Number : 70590-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128988

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128988
Availability details for
Quantity
  • Availability
  • Add to Cart