ENA394767607 Enamine

methyl 2-(2,3-dichlorophenyl)-2-hydroxyacetate

  • MDL number MFCD16693808
  • Cas Number : 1247786-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128990

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128990
Availability details for
Quantity
  • Availability
  • Add to Cart