ENA394767616 Enamine

N-(carbamothioylamino)-2,3-dihydro-1,4-benzodioxine-6-carboxamide

  • MDL number MFCD21493903
  • Cas Number : 1443980-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129002
Availability details for
Quantity
  • Availability
  • Add to Cart