ENA394767619 Enamine

3-cyclopentyl-3-oxopropanenitrile

  • MDL number MFCD04114391
  • Cas Number : 95882-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129015

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129015
Availability details for
Quantity
  • Availability
  • Add to Cart