ENA394767624 Enamine

3-(2,3-dihydro-1,4-benzodioxin-6-yl)-1H-1,2,4-triazole-5-thiol

  • MDL number MFCD11212860
  • Cas Number : 1094769-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 283...285
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129020

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129020
Availability details for
Quantity
  • Availability
  • Add to Cart