ENA394767631 Enamine

1-[1-(2-methanesulfonylethyl)-3,5-dimethyl-1H-pyrazol-4-yl]propan-1-one

  • MDL number MFCD18014926
  • Cas Number : 1282827-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155...157
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129049

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129049
Availability details for
Quantity
  • Availability
  • Add to Cart