ENA394767632 Enamine

1-(2-methanesulfonylethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD18889225
  • Cas Number : 1341874-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129051

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129051
Availability details for
Quantity
  • Availability
  • Add to Cart