ENA394767650 Enamine

1-(hydroxymethyl)cyclopentane-1-carbonitrile

  • MDL number MFCD19103512
  • Cas Number : 911060-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129076

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129076
Availability details for
Quantity
  • Availability
  • Add to Cart