ENA394767655 Enamine

ethyl 2-(1H-imidazol-1-yl)pyridine-3-carboxylate

  • MDL number MFCD19011643
  • Cas Number : 1443980-73-1
  • Molecular Weight : 217.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129085

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129085
Availability details for
Quantity
  • Availability
  • Add to Cart