ENA394767664 Enamine

4-(3,4-dimethylphenyl)-1,2-dihydrophthalazin-1-one

  • MDL number MFCD00278216
  • Cas Number : 93517-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 252...254
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129102

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129102
Availability details for
Quantity
  • Availability
  • Add to Cart