ENA394767666 Enamine

4-(4-methoxyphenyl)-2,2-dimethyl-4-oxobutanoic acid

  • MDL number MFCD01320131
  • Cas Number : 15118-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129104
Availability details for
Quantity
  • Availability
  • Add to Cart