ENA394767670 Enamine

(1R)-1-phenylpentan-1-amine hydrochloride

  • MDL number MFCD24842933
  • Cas Number : 529512-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129116
Availability details for
Quantity
  • Availability
  • Add to Cart