ENA394767671 Enamine

2,3-dimethylpent-2-enoic acid

  • MDL number MFCD24842934
  • Cas Number : 122630-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129117

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129117
Availability details for
Quantity
  • Availability
  • Add to Cart