ENA394767676 Enamine

2-methyl-5-(trifluoromethyl)piperidine

  • MDL number MFCD20311914
  • Cas Number : 1344335-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129123
Availability details for
Quantity
  • Availability
  • Add to Cart