ENA394767680 Enamine

2-(2,3-difluorophenyl)ethan-1-ol

  • MDL number MFCD01862011
  • Cas Number : 126163-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129162

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129162
Availability details for
Quantity
  • Availability
  • Add to Cart