ENA394767682 Enamine

3,5-dimethylpyridine-2-carboxylic acid

  • MDL number MFCD09839172
  • Cas Number : 4733-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151...153
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129172

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129172
Availability details for
Quantity
  • Availability
  • Add to Cart