ENA394767688 Enamine

2,2,2-trifluoro-N-(2-methylpyridin-4-yl)acetamide

  • MDL number MFCD21643709
  • Cas Number : 1443981-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129186
Availability details for
Quantity
  • Availability
  • Add to Cart