ENA394767693 Enamine

3-methoxy-2-methyl-4H-pyran-4-one

  • MDL number MFCD14553191
  • Cas Number : 4780-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129191

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129191
Availability details for
Quantity
  • Availability
  • Add to Cart