ENA394767697 Enamine

N-(1-cyano-1-methylethyl)-2,2,2-trifluoro-N-methylacetamide

  • MDL number MFCD24842939
  • Cas Number : 1443981-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129195

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129195
Availability details for
Quantity
  • Availability
  • Add to Cart