ENA394767699 Enamine

3-(2-nitrophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD11203721
  • Cas Number : 958891-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129198

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129198
Availability details for
Quantity
  • Availability
  • Add to Cart