ENA394767700 Enamine

N,2,3-triphenyl-2,5-dihydro-1,2,4-thiadiazol-5-imine

  • Cas Number : 69909-49-5
  • Molecular Weight : 329.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-((5E)-2,3-diphenyl-1,2,4-thiadiazol-5(2H)-ylidene)aniline
Inchi Code 1S/C20H15N3S/c1-4-10-16(11-5-1)19-22-20(21-17-12-6-2-7-13-17)24-23(19)18-14-8-3-9-15-18/h1-15H/b21-20+
Inchi Key MWKGMZRDURKJEZ-QZQOTICOSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129199
Availability details for
Quantity
  • Availability
  • Add to Cart