ENA394767709 Enamine

3,4-dihydro-1H-2-benzopyran-1-carboxamide

  • MDL number MFCD12815828
  • Cas Number : 860368-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129213

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129213
Availability details for
Quantity
  • Availability
  • Add to Cart