ENA394767710 Enamine

ethyl 4-(adamantan-1-yl)-2,4-dioxobutanoate

  • MDL number MFCD24842941
  • Cas Number : 847955-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129214

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129214
Availability details for
Quantity
  • Availability
  • Add to Cart