ENA394767726 Enamine

5-methoxy-4-(2-methylbutoxy)-2-nitrobenzaldehyde

  • MDL number MFCD17439984
  • Cas Number : 1443980-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129270
Availability details for
Quantity
  • Availability
  • Add to Cart