ENA394767731 Enamine

2-[(2-bromophenyl)methyl]propanedioic acid

  • MDL number MFCD03003303
  • Cas Number : 58380-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129292
Availability details for
Quantity
  • Availability
  • Add to Cart