ENA394767736 Enamine

6-(sulfanylmethyl)pyridine-2-carboxamide

  • MDL number MFCD20718771
  • Cas Number : 1379306-18-9
  • Molecular Weight : 168.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(sulfanylmethyl)-2-pyridinecarboxamide
Inchi Code 1S/C7H8N2OS/c8-7(10)6-3-1-2-5(4-11)9-6/h1-3,11H,4H2,(H2,8,10)
Inchi Key NEHIPPXPXVUHPH-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129308
Availability details for
Quantity
  • Availability
  • Add to Cart