ENA394767737 Enamine

tert-butyl 3-(2-fluorophenyl)-3-hydroxyazetidine-1-carboxylate

  • MDL number MFCD21596789
  • Cas Number : 1225439-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122073

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122073
Availability details for
Quantity
  • Availability
  • Add to Cart