ENA394767750 Enamine

1-chloro-4-(trimethylsilyl)but-3-yn-2-one

  • MDL number MFCD01862797
  • Cas Number : 18245-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129322
Availability details for
Quantity
  • Availability
  • Add to Cart