ENA394767751 Enamine

N-(2-methyl-1,3-benzothiazol-5-yl)-3-(trifluoromethyl)benzamide

  • MDL number MFCD06338751
  • Cas Number : 406194-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129324
Availability details for
Quantity
  • Availability
  • Add to Cart