ENA394767754 Enamine

ethyl 7-amino-1,3-dioxaindane-5-carboxylate

  • MDL number MFCD17276553
  • Cas Number : 1343291-63-3
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129328

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129328
Availability details for
Quantity
  • Availability
  • Add to Cart