ENA394767756 Enamine

8-methyl-8-azabicyclo[3.2.1]octane-3-carbonitrile

  • MDL number MFCD00970017
  • Cas Number : 96920-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129331

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129331
Availability details for
Quantity
  • Availability
  • Add to Cart