ENA394767769 Enamine

3-(propan-2-yl)-1-(2,2,2-trifluoroethyl)piperazine

  • MDL number MFCD20427031
  • Cas Number : 1443979-62-1
  • Molecular Weight : 210.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-isopropyl-1-(2,2,2-trifluoroethyl)piperazine
Inchi Code 1S/C9H17F3N2/c1-7(2)8-5-14(4-3-13-8)6-9(10,11)12/h7-8,13H,3-6H2,1-2H3
Inchi Key VCJOQYHIIZAOGU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129347
Availability details for
Quantity
  • Availability
  • Add to Cart