ENA394767771 Enamine

1-amino-4-(propan-2-yl)cyclohexane-1-carbonitrile

  • MDL number MFCD16853286
  • Cas Number : 804433-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129350

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129350
Availability details for
Quantity
  • Availability
  • Add to Cart