ENA394767774 Enamine

2-(4-bromophenyl)-3-methylbutanenitrile

  • MDL number MFCD11592218
  • Cas Number : 51632-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129355

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129355
Availability details for
Quantity
  • Availability
  • Add to Cart