ENA394767777 Enamine

4,6-dimethoxybenzene-1,3-disulfonyl dichloride

  • MDL number MFCD09033910
  • Cas Number : 80585-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129359
Availability details for
Quantity
  • Availability
  • Add to Cart