ENA394767789 Enamine

8-benzyl-3-oxa-1,8-diazaspiro[4.5]decan-2-one

  • MDL number MFCD18074228
  • Cas Number : 945892-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129376
Availability details for
Quantity
  • Availability
  • Add to Cart