ENA394767791 Enamine

[3-(adamantan-1-yl)-1H-pyrazol-5-yl]methanol

  • MDL number MFCD21567006
  • Cas Number : 493038-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129378

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129378
Availability details for
Quantity
  • Availability
  • Add to Cart