ENA394767801 Enamine

3-{1-[(4-methoxyphenyl)methyl]-1,2,3,6-tetrahydropyridin-4-yl}-1H-indole

  • MDL number MFCD24842969
  • Cas Number : 1352550-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129407
Availability details for
Quantity
  • Availability
  • Add to Cart