ENA394767809 Enamine

4-methoxypiperidine-1-carboximidamide hydrobromide

  • MDL number MFCD24842974
  • Cas Number : 1443980-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129428
Availability details for
Quantity
  • Availability
  • Add to Cart