ENA394767813 Enamine

2-[(thiophen-3-yl)methyl]propanedioic acid

  • MDL number MFCD03003280
  • Cas Number : 26415-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-140
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129443
Availability details for
Quantity
  • Availability
  • Add to Cart