ENA394767820 Enamine

1-methyl-5-(methylsulfanyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD00171475
  • Cas Number : 68700-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129451
Availability details for
Quantity
  • Availability
  • Add to Cart