ENA394767822 Enamine

2-[(3-fluorophenyl)methyl]-2-methylpropanedioic acid

  • MDL number MFCD24842978
  • Cas Number : 1443979-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129454
Availability details for
Quantity
  • Availability
  • Add to Cart