ENA394767823 Enamine

2-(ethylcarbamoyl)acetic acid

  • MDL number MFCD06208556
  • Cas Number : 773098-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129455
Availability details for
Quantity
  • Availability
  • Add to Cart