ENA394767910 Enamine

(2,3-dihydro-1H-inden-1-yl)methyl methanesulfonate

  • MDL number MFCD24842982
  • Cas Number : 209790-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129461
Availability details for
Quantity
  • Availability
  • Add to Cart