ENA394767911 Enamine

ethyl N-{[(4-bromophenyl)methyl]carbamothioyl}carbamate

  • MDL number MFCD24842983
  • Cas Number : 1443981-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129466

Safety Information

msds https://www.enaminestore.com/catalog/EN300-129466
Availability details for
Quantity
  • Availability
  • Add to Cart