ENA394767912 Enamine

2,5,5-trimethyl-2-(2-methyl-5-nitrophenyl)-1,3-dioxane

  • MDL number MFCD24842985
  • Cas Number : 217197-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129488

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129488
Availability details for
Quantity
  • Availability
  • Add to Cart