ENA394767923 Enamine

2-hydroxy-5-(methoxycarbonyl)pyridine-3-sulfonic acid

  • MDL number MFCD24843001
  • Cas Number : 1443980-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-129818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129818
Availability details for
Quantity
  • Availability
  • Add to Cart