ENA394767925 Enamine

2-amino-2,3-dihydro-1H-isoindol-1-one hydrochloride

  • MDL number MFCD24843004
  • Cas Number : 23990-36-5
  • Molecular Weight : 184.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-aminoisoindolin-1-one hydrochloride
Inchi Code 1S/C8H8N2O.ClH/c9-10-5-6-3-1-2-4-7(6)8(10)11;/h1-4H,5,9H2;1H
Inchi Key WVQAQTSNCMFWNG-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129836

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-129836
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart